missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15407776
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1001200-60-7
Molecular Formula C13H18BFO2
Molecular Weight (g/mol) 236.093
MDL Number MFCD18729904
InChI Key UNTVLIKQVDPHFS-UHFFFAOYSA-N
Synonym 2-4-fluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-3-methylphenylboronic acid pinacol ester, 4-fluoro-3-methylphenylboronic acid, pinacol ester, amtb633
PubChem CID 56776823
IUPAC Name 2-(4-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C

Specifications

Quantity 1 g
Formula Weight 236.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methylbenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'