missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylanisole, 98%, ACROS Organics™

Product Code. 10547763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547763 5 g Glass bottle 5g
10412261 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10547763 Supplier Thermo Scientific Acros Supplier No. 294350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Identificatori chimici

CAS 2338-54-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD00042293
InChI Key XZBXPBDJLUJLEU-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylanisole, 2-fluoro-5-methoxytoluene, 1-fluoro-4-methoxy-2-methyl-benzene, benzene, 1-fluoro-4-methoxy-2-methyl, pubchem2623, 4-fluoro-3-methylani, 4-fluoro-3-methylanisol, 3-methyl-4-fluoroanisole, 4-fluoro-3-methylanizole, acmc-1cr5e
PubChem CID 137546
IUPAC Name 1-fluoro-4-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)F

Specifica

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(CH3)OCH3
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
Quantity 5 g
Formula Weight 140.16
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-methylanisole, 98%
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.