missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-methylacetophenone, 97%, Thermo Scientific™

Product Code. 15485227
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485227 Supplier Thermo Scientific Alfa Aesar Supplier No. H32499.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4′-Fluoro-3′-methylacetophenone is used as pharmaceutical intermediate, and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-32-4
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD04972879
InChI Key SMSVMBMJEYTUOZ-UHFFFAOYSA-N
Synonym 1-4-fluoro-3-methylphenyl ethanone, 4-fluoro-3-methylacetophenone, 4'-fluoro-3'-methylacetophenone, 1-4-fluoro-3-methylphenyl ethan-1-one, 3-methyl-4-fluoroacetophenone, 1-acetyl-4-fluoro-3-methylbenzene, ethanone,1-4-fluoro-3-methylphenyl, ethanone, 1-4-fluoro-3-methylphenyl, pubchem19762, 4-fluor-3-methylacetophenone
PubChem CID 289328
IUPAC Name 1-(4-fluoro-3-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C(C)=C1

Specifications

Odor Characteristic
Refractive Index 1.5128
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-3'-methylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.