missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 15488965
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-92-3
Molecular Formula C9H5F4N
Molecular Weight (g/mol) 203.14
MDL Number MFCD22201068
InChI Key LHJDUSUXBSSXAY-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl-5-trifluoromethyl benzonitrile
PubChem CID 86277675
IUPAC Name 4-fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
SMILES CC1=C(C(=CC(=C1)C#N)C(F)(F)F)F

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 203.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'