missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxyphenylacetic acid, 95%, Thermo Scientific™

Product Code. 10567590
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10567590 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10567590 Supplier Thermo Scientific Maybridge Supplier No. MAY00208DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 946713-86-6
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C9H9FO3
Quantity 1 g
Synonym 4-fluoro-3-methoxyphenylacetic acid, 2-4-fluoro-3-methoxyphenyl acetic acid, 4-fluoro-3-methoxyphenyl acetic acid, ksc495a9p, 5-carboxymethyl-2-fluoroanisole, benzeneacetic acid, 4-fluoro-3-methoxy, 4-fluoro-3 methoxy phenyl acetic acid, 2-4-fluoranyl-3-methoxy-phenyl ethanoic acid
InChI Key YNNVYROHVVBVFI-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)CC(=O)O)F
IUPAC Name 2-(4-fluoro-3-methoxyphenyl)acetic acid
Molecular Weight (g/mol) 184.166
PubChem CID 26188830
Formula Weight 184.17
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-methoxyphenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.