missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. p-7047720
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11395485

Brand: Thermo Scientific Alfa Aesar L19673.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82846-18-2
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272134
InChI Key LWGCZCMLPRMKIZ-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973
PubChem CID 598436
IUPAC Name 4-fluoro-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)F

Specifications

Melting Point 206°C to 208°C
Quantity 1 g
Beilstein 4178071
Formula Weight 170.14
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.