missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-chloroaniline, TRC

Product Code. 30390578
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390578 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390578 Supplier Toronto Research Chemicals Supplier No. TRCF58901510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 367-21-5
Molecular Formula C6 H5 Cl F N
Quantity 10 g
Synonym Aniline, 3-chloro-4-fluoro- (6CI,7CI,8CI), 3-Chloro-4-fluorobenzenamine, 3-Chloro-4-fluoroaniline, 3-Chloro-4-fluorophenylamine, 4-Fluoro-3-chloroaniline, NSC 10290, m-Chloro-p-fluoroaniline
InChI Formula InChI=1S/C6H5ClFN/c7-5-3-4(9)1-2-6(5)8/h1-3H,9H2
SMILES Nc1ccc(F)c(Cl)c1
IUPAC Name 3-chloro-4-fluoroaniline
Molecular Weight (g/mol) 145.56
Formula Weight 145.0095
Chemical Name or Material 3-Chloro-4-fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.