missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11450551
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11450551

Brand: Thermo Scientific Alfa Aesar B20064.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-2-(trifluoromethyl)benzoyl chloride is used in medicine.

Solubility
Not miscible in water.

Notes
Store at 4°C. Store away from moisture and oxidizing agents under dry inert gas. Incompatible with active metals, bases and amines. Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189807-21-4
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.55
MDL Number MFCD00061158
InChI Key OEYHURRIOWWRMD-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-chlorocarbonyl-5-fluorobenzotrifluoride, benzoyl chloride, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, acmc-1bwn8, ksc540e2h, attercop-chm at107069, 2-trifluoromethyl-4-fluorobenzoylchlorid
PubChem CID 605680
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzoyl chloride
SMILES FC1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F

Specifications

Density 1.495
Boiling Point 157°C
Flash Point 85°C (185°F)
Refractive Index 1.468
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.