missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzonitrile, 98+%

Product Code. 11479037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479037 5 g 5g
11469037 1 g 1g
11384729 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479037 Supplier Thermo Scientific Alfa Aesar Supplier No. B20617.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 194853-86-6 | C8H3F4N | 189.11 g/mol

It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194853-86-6
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.11
MDL Number MFCD00061283
InChI Key LCCPQUYXMFXCAC-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-fluorobenzonitrile, 4-fluoro-2-trifluoromethyl-benzonitrile, 2-cyano-5-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolunitrile, benzonitrile, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzenecarbonitrile, pubchem4802
PubChem CID 605085
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzonitrile
SMILES FC1=CC=C(C#N)C(=C1)C(F)(F)F

Specifications

Melting Point 43°C to 45°C
Density 1.35
Boiling Point 194°C to 196°C
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water. Solubility in methanol is almost transparent.
Formula Weight 189.11
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.