missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 10638444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10638444 5 g Glass bottle 5g
10217420 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10638444 Supplier Thermo Scientific Acros Supplier No. 309600050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 141179-72-8
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040982
InChI Key JUHPDXOIGLHXTC-UHFFFAOYSA-M
Synonym 4-fluoro-2-trifluoromethyl benzoic acid, 4-fluoro-2 trifluoromethyl benzoic acid, alpha,alpha,alpha,4-tetrafluoro-o-toluic acid, benzoic acid, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzoicacid, pubchem4971, acmc-209clt, intermediates-zcf02077, ksc183o7t
PubChem CID 688255
SMILES [O-]C(=O)C1=CC=C(F)C=C1C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 124°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(CF3)COH
Quantity 5 g
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzoate
Formula Weight 208.11
Percent Purity ≥97.5%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 4-Fluoro-2(trifluoromethyl)benzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.