missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog Number p-3758037
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
141179-72-8
C8H3F4O2
207.10
MFCD00040982
JUHPDXOIGLHXTC-UHFFFAOYSA-M
4-fluoro-2-trifluoromethyl benzoic acid, 4-fluoro-2 trifluoromethyl benzoic acid, alpha,alpha,alpha,4-tetrafluoro-o-toluic acid, benzoic acid, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzoicacid, pubchem4971, acmc-209clt, intermediates-zcf02077, ksc183o7t
688255
[O-]C(=O)C1=CC=C(F)C=C1C(F)(F)F
This item is not returnable. View return policy
141179-72-8
C8H3F4O2
207.10
MFCD00040982
JUHPDXOIGLHXTC-UHFFFAOYSA-M
4-fluoro-2-trifluoromethyl benzoic acid, 4-fluoro-2 trifluoromethyl benzoic acid, alpha,alpha,alpha,4-tetrafluoro-o-toluic acid, benzoic acid, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzoicacid, pubchem4971, acmc-209clt, intermediates-zcf02077, ksc183o7t
688255
[O-]C(=O)C1=CC=C(F)C=C1C(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 124.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(CF3)CO2H
Quantity 1 g
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzoic acid
Formula Weight 208.11
Percent Purity 98%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 4-Fluoro-2(trifluoromethyl)benzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'