missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Brand:  Acros Organics 309600010

Code : 29

Additional Details : CAS Number : 141179-72-8 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10217420

  • £8.80 / 1g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Fluoro-2(trifluoromethyl)benzoic acid
97.5
98%
C8H4F4O2
208.11
141179-72-8
100.0
98%
FC6H3(CF3)CO2H
MFCD00040982

Specifications

4-Fluoro-2(trifluoromethyl)benzoic acid
121.0° to 124.0°C
Authentic
97.5
Glass bottle
98%
C8H4F4O2
208.11
Slightly yellow
1g
141179-72-8
100.0
Needle-Like Crystalline Powder
98%
FC6H3(CF3)CO2H
MFCD00040982
Videos
SDS