missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11488926
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
missing translation for 'unitSize':
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11488926 10 g 10g
11478926 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11488926 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A17067.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-39-5
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00007831
InChI Key LRCQLCWUUBSUOY-UHFFFAOYSA-N
PubChem CID 67853
IUPAC Name 4-fluoro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(F)C=C1C(F)(F)F

Specifications

Density 1.38
Boiling Point 70°C to 71°C (18 mmHg)
Flash Point 72°C (161°F)
Odor Characteristic
Refractive Index 1.463
Quantity 10 g
UN Number UN2810
Beilstein 2098758
Synonym 2-amino-5-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl aniline, 4-fluoro-2-trifluoromethyl-phenylamine, benzenamine, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluidine, alpha, 2-amino-5-fluoro benzotrifluoride, 4-fluoro-2-trifluoromethyl phenylamine, alpha,alpha,alpha-4-tetrafluoro-o-toluidine, benzenamine,4-fluoro-2-trifluoromethyl
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.