missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11478926
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11478926

Brand: Thermo Scientific Alfa Aesar A17067.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-39-5
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00007831
InChI Key LRCQLCWUUBSUOY-UHFFFAOYSA-N
PubChem CID 67853
IUPAC Name 4-fluoro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(F)C=C1C(F)(F)F

Specifications

Density 1.38
Boiling Point 70°C to 71°C (18 mmHg)
Flash Point 72°C (161°F)
Odor Characteristic
Refractive Index 1.463
Quantity 2 g
UN Number UN2810
Beilstein 2098758
Synonym 2-amino-5-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl aniline, 4-fluoro-2-trifluoromethyl-phenylamine, benzenamine, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluidine, alpha, 2-amino-5-fluoro benzotrifluoride, 4-fluoro-2-trifluoromethyl phenylamine, alpha,alpha,alpha-4-tetrafluoro-o-toluidine, benzenamine,4-fluoro-2-trifluoromethyl
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)aniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.