missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethoxy)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15487297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15487297 1 g 1g
15418905 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15487297

Brand: Thermo Scientific Alfa Aesar H34362.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1242252-68-1
Molecular Formula C8H6F4O2
Molecular Weight (g/mol) 210.128
MDL Number MFCD19687174
InChI Key QTZPHLXETXULFE-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethoxy benzyl alcohol, 4-fluoro-2-trifluoromethoxy phenyl methanol
PubChem CID 66523583
IUPAC Name [4-fluoro-2-(trifluoromethoxy)phenyl]methanol
SMILES C1=CC(=C(C=C1F)OC(F)(F)F)CO

Specifications

Quantity 1 g
Formula Weight 210.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-(trifluoromethoxy)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.