missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrotoluene, 98%, Thermo Scientific™

Product Code. 11490538
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490538 25 g 25g
11480538 5 g 5g
11400548 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490538 Supplier Thermo Scientific Alfa Aesar Supplier No. B24677.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-10-6
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007200
InChI Key SKWTUNAAJNDEIK-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
PubChem CID 67965
IUPAC Name 4-fluoro-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)F)[N+](=O)[O-]

Specifications

Melting Point 25°C to 27°C
Density 1.26
Boiling Point 137°C to 139°C (83 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.523
Quantity 25 g
UN Number UN2811
Beilstein 1946051
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitrotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.