missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrophenol, 99%, ACROS Organics™

Product Code. 10309410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10309410 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10309410 Supplier Thermo Scientific Acros Supplier No. 311970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 394-33-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 74°C to 77°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H4FNO3
Linear Formula FC6H3(NO2)OH
MDL Number MFCD00042528
Quantity 5 g
Synonym 2-nitro-4-fluorophenol, phenol, 4-fluoro-2-nitro, phenol,2-nitro,4-fluoro, 4-fluoro-2-nitro-phenol, 4-fluoronitrophenol, pubchem1503, acmc-1ah4y, ksc493s6p, 5-fluoro-2-hydroxynitrobenzene, 4-fluoro-2-nitrophenol
InChI Key ZHRLVDHMIJDWSS-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])O
IUPAC Name 4-fluoro-2-nitrophenol
Molecular Weight (g/mol) 157.1
PubChem CID 136236
Formula Weight 157.1
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitrophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.