missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrobenzonitrile, 97%

Product Code. 11384676
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384676 1 g 1g
11374676 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384676 Supplier Thermo Scientific Alfa Aesar Supplier No. H32610.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80517-21-1 | C7H3FN2O2 | 166.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80517-21-1
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.111
MDL Number MFCD00277447
InChI Key MNEAKKQYFSYZEU-UHFFFAOYSA-N
Synonym 2-cyano-5-fluoronitrobenzene, benzonitrile, 4-fluoro-2-nitro, buttpark 100\07-73, pubchem4795, acmc-209pjl, 4-fluoro-2-nitrobenzonitrle, ksc497q7l, 4-fluoro-2-nitro-benzonitrile, benzonitrile,4-fluoro-2-nitro, 4-fluoro-2-nitrobenzenecarbonitrile
PubChem CID 2774652
IUPAC Name 4-fluoro-2-nitrobenzonitrile
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])C#N

Specifications

Melting Point 69°C to 73°C
Quantity 1 g
UN Number UN3439
Formula Weight 166.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-nitrobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.