missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-nitroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
500 g
Description
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Melting Point | 90°C to 94°C |
Flash Point | 89°C (192°F) |
Quantity | 500 g |
Beilstein | 2210197 |
Solubility Information | Insoluble in water. |
Formula Weight | 156.12 |
Percent Purity | 98% |
Chemical Name or Material | 4-Fluoro-2-nitroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'