Learn More
4-Fluoro-2-nitroaniline, 98%
Description
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Specifications
| Melting Point | 90°C to 94°C |
| Flash Point | 89°C (192°F) |
| Quantity | 100 g |
| Beilstein | 2210197 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 156.12 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Fluoro-2-nitroaniline |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.