missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-nitroacetanilide, 99+%, Thermo Scientific™

Product Code. 10266480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266480 5 g Glass bottle 5g
10686524 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266480 Supplier Thermo Scientific Acros Supplier No. 249660050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 448-39-5
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.16
MDL Number MFCD00031517
InChI Key UZBZEUCQENVPQB-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-nitroacetanilide, n-4-fluoro-2-nitrophenyl acetamide, 4-fluoro-2-nitroacetanilide, maybridge1_003501, 4/'-fluoro-2/'-nitroacetanilide, labotest-bb lt00455473, labotest-bb lt03332301, acetamide,n-4-fluoro-2-nitrophenyl, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide #
PubChem CID 136289
IUPAC Name N-(4-fluoro-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Ochre to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3NO2NHC(O)CH3
Quantity 5 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in non-polar solvents,soluble in polar,solvents
Formula Weight 198.16
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Fluoro-2'-nitroacetanilide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoro-2'-nitroacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.