missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%

Product Code. 15482349
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482349 1 g 1g
15469256 250 mg 250mg
15492349 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482349 Supplier Thermo Scientific Alfa Aesar Supplier No. H64928.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 179062-05-6 | C7H4F4N2O2 | 224.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179062-05-6
Molecular Formula C7H4F4N2O2
Molecular Weight (g/mol) 224.115
MDL Number MFCD03094267
InChI Key WOSGMZUIHAFPSS-UHFFFAOYSA-N
Synonym 5-amino-2-fluoro-4-nitrobenzotrifluoride, 4-fluoro-2-nitro-5-trifluoromethyl aniline, benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl, pubchem4933, ksc106o9t, 2-fluoro-4-nitro-5-aminobenzotrifluoride, 3-amino-6-fluoro-4-nitrobenzotrifluoride, 4-fluoro-2-nitro-5-trifluoromethyl benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl phenylamine
PubChem CID 2737686
IUPAC Name 4-fluoro-2-nitro-5-(trifluoromethyl)aniline
SMILES C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F

Specifications

Color White
Quantity 1 g
UN Number UN2811
Beilstein 14360199
Formula Weight 224.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-nitro-5-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.