missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10611663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10611663 1 g Glass bottle 1g
10782833 5 g Glass bottle 5g
10003393 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10611663 Supplier Thermo Scientific Acros Supplier No. 367940010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 139911-29-8
MDL Number MFCD02093072
InChI Key IQMLIVUHMSIOQP-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp
PubChem CID 2734665
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)F)C)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H8BFO2
Quantity 1 g
Molecular Weight (g/mol) 153.947
Formula Weight 153.95
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.