missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzyl alcohol, 99%

Product Code. 15445087
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445087 5 g 5g
15435087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445087 Supplier Thermo Scientific Alfa Aesar Supplier No. H32364.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80141-91-9 | C8H9FO | 140.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80141-91-9
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD03701058
InChI Key YSULUXOLTMBSFF-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylphenyl methanol, 4-fluoro-2-methylbenzyl alcohol, 4-fluoro-2-methylbenzylalcohol, rarechem al bd 0506, 4-fluoro-2-methylbenzenemethanol, benzenemethanol, 4-fluoro-2-methyl, 4-fluoro-2-methyl-phenyl-methanol, 4-fluoro-2-methylphenyl methan-1-ol, 4'-fluoro-2-methylbenzyl alcohol
PubChem CID 3872017
IUPAC Name (4-fluoro-2-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)F)CO

Specifications

Quantity 5 g
Formula Weight 140.16
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.