missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10153144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153144 5 g Glass bottle 5g
10480694 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153144 Supplier Thermo Scientific Acros Supplier No. 399930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 147754-12-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD03095106
InChI Key BJBXUIUJKPOZLV-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l
PubChem CID 3826055
IUPAC Name 4-fluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)F)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)CN
Quantity 5 g
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzonitrile
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylbenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.