missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10686840
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 179899-07-1
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD03788423
InChI Key ADJBXDCXYMCCAD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-4-fluorophenylboronic acid, 4-fluoro-2-methoxybenzeneboronic acid, 4-fluoro-2-methoxy-phenyl boronic acid, 4-fluoro-2-methoxyphenyl-boronic acid, 5-fluoroanisole-2-boronic acid, 4-fluoro-2-methoxy phenyl boronic acid, boronic acid, 4-fluoro-2-methoxyphenyl, pubchem19716, acmc-1c2tm
PubChem CID 2782672
IUPAC Name (4-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC(F)=CC=C1B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 133.9°C to 137.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 169.95
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methoxyphenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'