missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodobenzoic acid, TRC

Product Code. 30390225
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390225 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390225 Supplier Toronto Research Chemicals Supplier No. TRCF5919981G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluoro-2-iodobenzoic acid
CAS 56096-89-0
Molecular Formula C7H4FIO2
Quantity 1 g
InChI Formula InChI=1S/C7H4FIO2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)
SMILES O=[13C]([13C](CC(C)C)(CC=C)[13C](N1)=O)N[13C]1=O
Molecular Weight (g/mol) 266.01
Product Title
Select an issue

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.