missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodobenzoic acid, 97%

Product Code. 15451828
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15451828

Brand: Thermo Scientific Alfa Aesar H55531.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56096-89-0 | C7H4FIO2 | 266.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56096-89-0
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD04108278
InChI Key DUKFTVLJAXWGPI-UHFFFAOYSA-N
Synonym 4-fluoro-2-iodo-benzoic acid, 2-iodo-4-fluorobenzoic acid, 4-fluoro-2-iodobenzoicacid, benzoic acid, 4-fluoro-2-iodo, benzoic acid, 4-fluoroiodo, pubchem1389, acmc-1atr7, ksc494a9j, benzoic acid,4-fluoro-2-iodo, buttpark 83\07-33
PubChem CID 12520164
IUPAC Name 4-fluoro-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1F)I)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-iodobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.