missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,6-dimethylaniline, 97%

Product Code. 11451517
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451517 1 g 1g
11461517 5 g 5g
11471517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451517 Supplier Thermo Scientific Alfa Aesar Supplier No. B24965.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 392-70-1 | C8H10FN | 139.173 g/mol

4-Fluoro-2,6-dimethylaniline is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,6-dimethylaniline is used as an intermediate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-70-1
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03792669
InChI Key MPNDLCBMLBACDD-UHFFFAOYSA-N
Synonym 4-fluoro-2,6-dimethylphenylamine, 2,6-dimethyl-4-fluoroaniline, acmc-20amil, pubchem4426, 4-fluoro-2,6-dimethyl-aniline, 4-fluoranyl-2,6-dimethyl-aniline, benzenamine,4-fluoro-2,6-dimethyl
PubChem CID 1403908
IUPAC Name 4-fluoro-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)F

Specifications

Quantity 1 g
Refractive Index 1.5345
Formula Weight 139.17
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2,6-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.