missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-1-naphthoic Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 4-Fluoro-1-naphthoic Acid |
| CAS | 573-03-5 |
| Molecular Formula | C11H7O2F |
| Quantity | 50 mg |
| Synonym | NSC 10831, 4-Fluoro-1-naphthalenecarboxylic Acid |
| InChI Formula | InChI=1S/C11H7FO2/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H,(H,13,14) |
| SMILES | OC(=O)c1ccc(F)c2ccccc12 |
| IUPAC Name | 4-fluoronaphthalene-1-carboxylic acid |
| Molecular Weight (g/mol) | 190.17 |
| Formula Weight | 190.043 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction