missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-naphthoic acid, 97%

Product Code. 11402808
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11402808

Brand: Thermo Scientific Alfa Aesar A16778.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 573-03-5 | C11H7FO2 | 190.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-03-5
Molecular Formula C11H7FO2
Molecular Weight (g/mol) 190.173
MDL Number MFCD00004011
InChI Key DEWIOKQDRWFLFW-UHFFFAOYSA-N
Synonym 4-fluoro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 4-fluoro, 4-fluoronaphthalenecarboxylic acid, 4-fluoro-1-naphthalenecarboxylic acid, 4-fluoro-1-naphthalene carboxylic acid, 4-fluoro-naphthalene-1-carboxylic acid, acmc-1atch, maybridge3_003028, ksc270s4j, 1-carboxy-4-fluoronaphthalene
PubChem CID 68448
IUPAC Name 4-fluoronaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=C2F)C(=O)O

Specifications

Melting Point 223°C to 226°C
Quantity 1 g
Beilstein 2257292
Formula Weight 190.18
Percent Purity 97%
Chemical Name or Material 4-Fluoro-1-naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.