missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-indanone, 97%

Product Code. 15465638
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465638 5 g 5g
15455638 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465638 Supplier Thermo Scientific Alfa Aesar Supplier No. H59356.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 699-99-0 | C9H7FO | 150.152 g/mol

It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

Solubility
Soluble in alcohol and chloroform

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-99-0
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.152
MDL Number MFCD00797829
InChI Key HOMSJDBZHCPYHY-UHFFFAOYSA-N
Synonym 4-fluoro-1-indanone, 4-fluoro-2,3-dihydro-1h-inden-1-one, 4-fluoro-indan-1-one, 4-fluoroindan-1-one, 1h-inden-1-one, 4-fluoro-2,3-dihydro, 4-fluoro-2,3-dihydro-1-indenone, 4-fluoroindane-1-one, acmc-20a8cy, 4-fluoranyl-2,3-dihydroinden-1-one
PubChem CID 12174986
IUPAC Name 4-fluoro-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)F

Specifications

Melting Point 72°C to 76°C
Quantity 5 g
Solubility Information Soluble in alcohol and chloroform
Formula Weight 150.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-1-indanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.