missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose palmitate, 90%

Product Code. 15478059
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 15478059

Brand: Thermo Scientific Chemicals J66627.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26446-38-8 | C28H52O12

Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMSO, DMF, ethanol and water

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26446-38-8
Molecular Formula C28H52O12
MDL Number MFCD00047545
Synonym beta-D-Fructofuranosyl-alpha-D-glucopyranoside monohexadecanoate

Specifications

Color White
Quantity 500 g
Solubility Information Soluble in DMSO, DMF, ethanol and water
Formula Weight 508.72
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Sucrose palmitate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.