missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,2-dimethoxybenzene, 98%

Product Code. 11429066
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429066 25 g 25g
11419066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429066 Supplier Thermo Scientific Alfa Aesar Supplier No. A17936.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 398-62-9 | C8H9FO2 | 156.156 g/mol

4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

Solubility
Soluble in water 0.079 g/L 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 398-62-9
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00012201
InChI Key DAGKHJDZYJFWSO-UHFFFAOYSA-N
Synonym 4-fluoroveratrole, 1,2-dimethoxy-4-fluorobenzene, 3,4-dimethoxyfluorobenzene, 1-fluoro-3,4-dimethoxybenzene, 4-fluoro-1,2-dimethoxy-benzene, 3,4-dimethoxy-1-fluorobenzene, 4-fluoroveratrol, pubchem3048, acmc-1acnf, fluorobenzene, 3,4-methoxy
PubChem CID 593640
IUPAC Name 4-fluoro-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)F)OC

Specifications

Density 1.171
Boiling Point 120°C to 123°C (45 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.51
Quantity 25 g
Beilstein 2441600
Solubility Information Soluble in water 0.079g/L 25°C.
Formula Weight 156.16
Percent Purity 98%
Chemical Name or Material 4-Fluoro-1,2-dimethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.