missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro(1,1'-biphenyl)-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10453963
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453963 1 g Amber Glass Bottle 1g
10639534 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453963 Supplier Thermo Scientific Maybridge Supplier No. MO07015DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116748-66-4
Molecular Formula C12H8ClFO2S
Molecular Weight (g/mol) 270.702
MDL Number MFCD01631921
InChI Key CIDMHDJTWVMBIF-UHFFFAOYSA-N
Synonym 4'-fluorobiphenyl-4-sulfonyl chloride, 4-4-fluorophenyl benzenesulfonyl chloride, 4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride, 4'-fluoro-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro, 4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride, acmc-20apun, 4'-fluorobiphenyl-4-sulfonylchloride, 4'-fluoro-4-biphenylsulfonyl chloride
PubChem CID 2794749
IUPAC Name 4-(4-fluorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 270.71
Percent Purity 97%
Chemical Name or Material 4';-Fluoro[1, 1';-biphenyl]-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.