missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylpyridine hydrochloride, 97%

Product Code. 15420288
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420288 1 g 1g
15430288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420288 Supplier Thermo Scientific Alfa Aesar Supplier No. H53455.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352530-29-1 | C7H6ClN | 139.582 g/mol

4-pyridylacetylene hydrochloride is used in the preparation of alkynyl platinum(II) compounds with dichloro (1,2-bis(diphenylphosphino)ethane)platinium(II) complex in the presence of base and copper iodide as catalyst. It is used as an intermediate in the synthesis of 1,4-bis(4-pyridyl)butadiyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-pyridylacetylene hydrochloride is used in the preparation of alkynyl platinum(II) compounds with dichloro (1,2-bis(diphenylphosphino)ethane)platinium(II) complex in the presence of base and copper iodide as catalyst. It is used as an intermediate in the synthesis of 1,4-bis(4-pyridyl)butadiyne.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352530-29-1
Molecular Formula C7H6ClN
Molecular Weight (g/mol) 139.582
MDL Number MFCD03093912
InChI Key KFBZWZGIZHLUBX-UHFFFAOYSA-N
Synonym 4-ethynylpyridine hydrochloride, 4-ethynylpyridine hcl, pyridine, 4-ethynyl-, hydrochloride, 4-ethynylpyridinehychloride salt, 4-ethynylpyridinehydrochloride, pubchem19527, 4-pyridylacetylene hydrochloride, 4-ethynyl pyridine hydrochloride, 4-ethynyl-pyridine hydrochloride, 4-ethynylpyridine hydrochloride 1:1
PubChem CID 16216931
IUPAC Name 4-ethynylpyridine;hydrochloride
SMILES C#CC1=CC=NC=C1.Cl

Specifications

Melting Point ∽150°C (decomposition)
Quantity 1 g
Formula Weight 139.58
Percent Purity 97%
Chemical Name or Material 4-Ethynylpyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.