missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylbenzonitrile, 97%

Product Code. 15487748
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487748 5 g 5g
15477748 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487748 Supplier Thermo Scientific Alfa Aesar Supplier No. H61244.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3032-92-6 | C9H5N | 127.146 g/mol

4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids and organic materials.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 3032-92-6
Molecular Formula C9H5N
Molecular Weight (g/mol) 127.146
MDL Number MFCD04974058
InChI Key LAGNMUUUMQJXBF-UHFFFAOYSA-N
Synonym 4-ethynyl-benzonitrile, 4-cyanophenylacetylene, benzonitrile, 4-ethynyl, p-cyanophenylacetylene, 4-ethynyl-bezonitrile, benzonitrile,4-ethynyl, 4-ethynylbenzenecarbonitrile, 4-ethynylbenzonitrile, ksc564s8b, benzonitrile, 4-ethynyl-9ci
PubChem CID 4547245
IUPAC Name 4-ethynylbenzonitrile
SMILES C#CC1=CC=C(C=C1)C#N

Spezifikation

Melting Point 156°C to 160°C
Color Yellow
Quantity 5 g
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 127.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Ethynylbenzonitrile
compliance-icons
Produktidentifikator
  • 4-Ethynylbenzonitrile
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 4-Ethynylbenzonitrile

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.