missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyltoluene, 97%

Product Code. 11331517
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331517 5 g 5g
11341517 25 g 25g
11351517 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331517 Supplier Thermo Scientific Alfa Aesar Supplier No. L02120.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-96-8 | C9H12 | 120.195 g/mol

4-Ethyltoluene is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethyltoluene is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-96-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009263
InChI Key JRLPEMVDPFPYPJ-UHFFFAOYSA-N
Synonym 4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
PubChem CID 12160
IUPAC Name 1-ethyl-4-methylbenzene
SMILES CCC1=CC=C(C=C1)C

Specifications

Melting Point -62°C
Density 0.861
Boiling Point 161°C to 163°C
Flash Point 43°C (109°F)
Refractive Index 1.495
Quantity 5 g
UN Number UN3295
Beilstein 1903007
Solubility Information Insoluble in water.
Formula Weight 120.2
Percent Purity 97%
Chemical Name or Material 4-Ethyltoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.