missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylthiophenol, 97%

Product Code. 11455987
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455987 1 g 1g
11465987 5 g 5g
11475987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455987 Supplier Thermo Scientific Alfa Aesar Supplier No. B25026.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4946-13-8 | C8H9S | 137.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4946-13-8
Molecular Formula C8H9S
Molecular Weight (g/mol) 137.22
MDL Number MFCD00040229
InChI Key WWQQPHUHTAZWDH-UHFFFAOYSA-M
Synonym 4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, acmc-209kg5
PubChem CID 2758914
IUPAC Name 4-ethylbenzenethiol
SMILES CCC1=CC=C([S-])C=C1

Specifications

Boiling Point 119°C to 120°C (30 mmHg)
Odor Stench
Refractive Index 1.5645
Quantity 1 g
UN Number UN2810
Beilstein 1905734
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 4-Ethylthiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.