missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylthio)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11378566
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378566 1 g 1g
11388566 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378566 Supplier Thermo Scientific Alfa Aesar Supplier No. L17623.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145349-76-4
Molecular Formula C8H11BO2S
Molecular Weight (g/mol) 182.044
MDL Number MFCD01318146
InChI Key LMUASBOUSAMURX-UHFFFAOYSA-N
Synonym 4-ethylthiophenylboronic acid, 4-ethylthio benzeneboronic acid, 4-ethylthio phenylboronic acid, 4-ethylsulfanyl phenyl boronic acid, 4-ethylmercapto benzeneboronic acid, 4-ethylthiobenzeneboronic acid, 4-ethylsulfanyl phenylboronic acid, p-ethylthio phenylboronic acid, boronic acid, 4-ethylthio phenyl, 4-ethylsulfanyl benzeneboronic acid
PubChem CID 2734353
IUPAC Name (4-ethylsulfanylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)SCC)(O)O

Specifications

Melting Point 132°C to 136°C
Quantity 1 g
Beilstein 8763159
Formula Weight 182.04
Percent Purity 98%
Chemical Name or Material 4-(Ethylthio)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.