missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylresorcinol, 98%

Product Code. p-7025866
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11467174

Brand: Thermo Scientific Alfa Aesar B21575.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2896-60-8 | C8H10O2 | 138.166 g/mol

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Solubility in methanol; almost transparency. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2896-60-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002283
InChI Key VGMJYYDKPUPTID-UHFFFAOYSA-N
Synonym 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol
PubChem CID 17927
IUPAC Name 4-ethylbenzene-1,3-diol
SMILES CCC1=C(C=C(C=C1)O)O

Specifications

Melting Point 95°C to 98°C
Quantity 5 g
Beilstein 1238231
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Ethylresorcinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.