missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylresorcinol, 98%

Product Code. 10182620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182620 25 g Glass bottle 25g
10265200 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182620 Supplier Thermo Scientific Acros Supplier No. 118950250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2896-60-8 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2896-60-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002283
InChI Key VGMJYYDKPUPTID-UHFFFAOYSA-N
Synonym 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol
PubChem CID 17927
IUPAC Name 4-ethylbenzene-1,3-diol
SMILES CCC1=C(C=C(C=C1)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 99.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 06, I, 441
Formula Weight 138.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethylresorcinol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylresorcinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.