missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylpyridine, 98%

Product Code. 11475323
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475323 250 g 250g
11465323 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475323 Supplier Thermo Scientific Alfa Aesar Supplier No. A12883.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 536-75-4 | C7H9N | 107.156 g/mol

4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

Solubility
Slightly soluble in water (34 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 536-75-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006450
InChI Key VJXRKZJMGVSXPX-UHFFFAOYSA-N
Synonym pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine
PubChem CID 10822
IUPAC Name 4-ethylpyridine
SMILES CCC1=CC=NC=C1

Spécification

Melting Point -91°C
Density 0.942
pH 10
Boiling Point 167°C to 168°C
Flash Point 52°C (126°F)
Odor Unpleasant
Refractive Index 1.501
Quantity 250 g
UN Number UN1993
Beilstein 106481
Merck Index 14,3849
Solubility Information Slightly soluble in water (34g/L at 20°C).
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material 4-Ethylpyridine
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.