missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate (Ethylpropyl Analogue of Oxybutynin), Mikromol™

Product Code. 17943356
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943356 Supplier Mikromol Supplier No. MM0047.07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate (Ethylpropyl Analogue of Oxybutynin)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate
CAS 1215677-72-7
Analyte or Component Names 4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate
Shipping Condition Room Temperature
Molecular Formula C23 H33 N O3
Certifications/Compliance ISO 17025
Synonym Oxybutynin Hydrochloride Imp. E (EP), Oxybutynin Imp. E (EP), 4-(Ethylpropylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate, Ethylpropyl Analogue of Oxybutynin, Oxybutynin Hydrochloride Impurity E, Oxybutynin Impurity E
InChI Formula InChI=1S/C47H53ClN3O3.BrH/c1-49(2)43(52)46(38-17-9-5-10-18-38,39-19-11-6-12-20-39)31-35-51(33-29-45(54,30-34-51)37-25-27-42(48)28-26-37)36-32-47(44(53)50(3)4,40-21-13-7-14-22-40)41-23-15-8-16-24-41;/h5-28,54H,29-36H2,1-4H3;1H/q+1;/p-3
SMILES CCCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
IUPAC Name 4-[ethyl(propyl)amino]but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate
Molecular Weight (g/mol) 371.51
Quantity 100 mg
Formula Weight 371.246 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.