missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10301184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301184 5 g Glass bottle 5g
10773203 1 g Glass bottle 1g
10530594 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10301184 Supplier Thermo Scientific Acros Supplier No. 359320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 63139-21-9
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD00859377
InChI Key RZCPLOMUUCFPQA-UHFFFAOYSA-N
Synonym 4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
PubChem CID 2734352
IUPAC Name (4-ethylphenyl)boronic acid
SMILES CCC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,methanol
Formula Weight 149.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.