missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl-d4 Sulfate Potassium Salt, TRC

Product Code. 30386372
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386372 25 mg -
30371017 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386372 Supplier Toronto Research Chemicals Supplier No. TRCE92586725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8 D4 H5 O4 S . K
Molecular Weight (g/mol) 244.343
Synonym 4-Ethylphenol-d4 1-(Hydrogen sulfate) Potassium Salt
IUPAC Name potassium;(2,3,5,6-tetradeuterio-4-ethyl-phenyl) sulfate
SMILES [K+].[2H]c1c([2H])c(OS(=O)(=O)[O-])c([2H])c([2H])c1CC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H10O4S.K/c1-2-7-3-5-8(6-4-7)12-13(9,10)11;/h3-6H,2H2,1H3,(H,9,10,11);/q;+1/p-1/i3D,4D,5D,6D;
Formula Weight 244.011
Chemical Name or Material 4-Ethylphenyl-d4 Sulfate Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.