missing translation for 'onlineSavingsMsg'
Learn More

(4-Ethylphenyl)acetic Acid, TRC

Product Code. 30386641
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386641 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386641 Supplier Toronto Research Chemicals Supplier No. TRCE925820100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Ethylphenyl)acetic Acid
CAS 14387-10-1
Molecular Formula C10H12O2
Quantity 100 mg
Synonym 2-(4-Ethylphenyl)acetic Acid, 4-Ethylphenylacetic Acid, p-Ethylphenylacetic Acid
InChI Formula InChI=1S/C10H12O2/c1-2-8-3-5-9(6-4-8)7-10(11)12/h3-6H,2,7H2,1H3,(H,11,12)
SMILES O=C(O)CC1=CC=C(C=C1)CC
IUPAC Name 2-(4-ethylphenyl)acetic acid
Molecular Weight (g/mol) 164.2
Formula Weight 164.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.