missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenol, 97%, Thermo Scientific Chemicals

Catalog Number 11455407
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A receptor agonists endowed with antinociceptive activity. It is also used in the preparation of isopropylamides as potent and noncovalent proteosome inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A recep
TRUSTED_SUSTAINABILITY
CAS 123-07-9
Melting Point 40°C to 45°C
Density 1.011
Boiling Point 218°C to 219°C
Flash Point 100°C (212°F)
Molecular Formula C8H10O
Refractive Index 1.533
MDL Number MFCD00002393
Quantity 250 g
UN Number UN2430
Beilstein 1363317
Synonym p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
Solubility Information Soluble in water. (4.9g/L) at 25°C
InChI Key HXDOZKJGKXYMEW-UHFFFAOYSA-N
SMILES CCC1=CC=C(C=C1)O
IUPAC Name 4-ethylphenol
Molecular Weight (g/mol) 122.167
PubChem CID 31242
ChEBI CHEBI:49584
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 4-Ethylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'