missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylmorpholine, Mikromol™

Product Code. 30049349
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049349 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049349 Supplier Mikromol Supplier No. MM3771.030100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Ethylmorpholine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Ethylmorpholine
CAS 100-74-3
Analyte or Component Names 4-Ethylmorpholine
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O
Certifications/Compliance ISO 9001
Synonym 4-Ethylmorpholine, Dabco NEM, Ethylmorpholine, Jeffcat NEM, KL 22, Kaolizer 22, N-Ethylmorpholine, NEM, NSC 6110, Toyocat NEM
InChI Formula InChI=1S/C6H13NO/c1-2-7-3-5-8-6-4-7/h2-6H2,1H3
SMILES CCN1CCOCC1
IUPAC Name 4-ethylmorpholine
Molecular Weight (g/mol) 115.174
Quantity 100 mg
Formula Weight 115.0997 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.