missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcatechol, TRC

Product Code. 30388491
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388491 5 g -
30373570 100 mg 100mg
30369397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388491 Supplier Toronto Research Chemicals Supplier No. TRCE9017605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-39-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.16
Synonym 1,2-Benzenediol, 4-ethyl-, Pyrocatechol, 4-ethyl- (6CI,8CI), 4-Ethyl-1,2-benzenediol, 1-Ethyl-3,4-benzenediol, 1-Ethyl-3,4-dihydroxybenzene, 4-Ethyl-1,2-dihydroxybenzene, 4-Ethyl-2-hydroxyphenol, 4-Ethylcatechol, 4-Ethylpyrocatechol, Ethylcatechol, Ethylpyrocatechol, 4-Ethylbenzene-1,2-diol
IUPAC Name 4-ethylbenzene-1,2-diol
SMILES CCc1ccc(O)c(O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H10O2/c1-2-6-3-4-7(9)8(10)5-6/h3-5,9-10H,2H2,1H3
Formula Weight 138.0681
Chemical Name or Material 4-Ethylcatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.